Z742615
PAGE-Optimizer™ Sample Preparation Column
10 columns
About This Item
Code UNSPSC :
41123100
Nomenclature NACRES :
NA.56
Produits recommandés
Caractéristiques
10 columns
Conditionnement
pkg of 10 ea
Fabricant/nom de marque
G-BioSciences 786-947
Température de stockage
2-8°C
Description générale
PAGE-Optimizer™ is a unique, spin column for the preparation of samples for SDS-PAGE. The columns contain a proprietary separation matrix that removes contaminants such as salts, detergents, cellular agents and other common buffering agents. PAGE-Optimizer™ helps eliminate smeared lanes, distorted and/ or smiling bands and swollen lanes. PAGE-Optimizer™ has also been shown to reveal protein bands previously masked by the above interfering agents.
Include: 10 columns
Include: 10 columns
Informations légales
PAGE-Optimizer is a trademark of Geno Technology, Inc.