Principaux documents
TMO4115-2000
Nalgene® sterile, single-use Erlenmeyer flask with vented closure
PETG, bottle volume 2000 mL, case of 4 ea
Synonyme(s) :
Nalgene Erlenmeyer flask, single use plastic flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.13
Matériaux
PETG
Stérilité
sterile
Caractéristiques
vented
Conditionnement
case of 4 ea
Fabricant/nom de marque
Nalgene 4115-2000
Volume du flacon
2000 mL
Description générale
Save preparation time and avoid contamination risk by using the Nalgene® Single-Use PETG Erlenmeyer Flasks, which are available with a flat or baffled bottom. These flasks are ideal for suspension of cell culture, media preparation, mixing and storage. Crystal-clear PETG is robust and break resistant, increasing personal safety. All sizes are available with a vented or non-vented closure.
Caractéristiques et avantages
  • Sterile and single us, reducing contamination risk
  • BPA-free
  • Collapses when autoclaved, reducing bio hazardous waste
  • Graduated for quick volume assessment
  • Increases user safety due to the break-resistant and robust design of the crystal-clear PETG material
  • Facilitates visual inspection and monitoring of contents due to its crystal-clear composition
  • Equipped with a vented cap to support gas exchange during processes like cell culture
Autres remarques
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries