Principaux documents
OH30456390
Ohaus® Navigator® Portable Balance
Model NVT4201, weighing capacity 4200 g, resolution: 0.1 g, AC/DC input 230 V, universal plug set
About This Item
Code UNSPSC :
41111502
Nomenclature NACRES :
NB.71
Matériaux
metal weighing plate
Caractéristiques
CE compliant
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30456390
Plage de mesure de la capacité de pesée
4200 g, resolution: 0.1 g
weighing pan W × L
17.4 cm × 23 cm
Description générale
OHAUS raises the bar in price-performance ratio with the newly-designed Navigator series which is perfect for general weighing applications in laboratory, industrial and education settings. Ruggedly constructed and multifunctional, the Navigator′s a powerful balance that can handle a diverse range of weighing applications with features such as fast stabilization time, overload protection at four times its rated capacity, and easy operation.
Application
Weighing, Parts Counting, Percent Weighing, Checkweighing
Caractéristiques et avantages
Menu and calibration lock switch, mechanical and software overload/underload protection, stability indicator, low battery indicator, auto shut-off, user-selectable print options, user-selectable communication settings
Informations légales
Navigator is a registered trademark of Ohaus Corp.
Ohaus is a registered trademark of Ohaus Corp.