Principaux documents
OH30253072
Ohaus® Scout® Portable Balance
Model SKX422, weighing capacity 420 g, resolution: 0.01 g, AC/DC input 230 V, universal plug set
About This Item
Code UNSPSC :
41111502
Nomenclature NACRES :
NB.71
Matériaux
metal weighing plate
Caractéristiques
CE compliant
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 30253072
Plage de mesure de la capacité de pesée
420 g, resolution: 0.01 g
Diam. de la nacelle de pesée
12 cm
Description générale
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Application
Weighing, Parts Counting, Percent Weighing, Check Weighing, Totalization, Display Hold
Caractéristiques et avantages
Stability indicator, stackable design with optional storage cover (excluding 1mg models), mechanical and software overload/underload protection, low battery indicator, auto shut-off, auto tare, span calibration mass included on models up to 620g capacity
Informations légales
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.