Principaux documents
HS24490M
Lab Alert® triple timer and clock
About This Item
Code UNSPSC :
41122411
Nomenclature NACRES :
NB.51
Fabricant/nom de marque
Heathrow Scientific HS24490M
Caractéristiques et avantages
  • Three channels to count up/down with
  • Simultaneous display, time-out, and 60 second audible alarm
  • Channels may be started independently or simultaneously
  • Timer automatically counts
  • 86 x 66 x 22 mm
Informations légales
Lab Alert is a registered trademark of Heathrow Scientific