Merck
GE18-3585-01
Filter Kit, HR 16
Synonyme(s) :
18-3585-01, Cytiva column accessory, accessory, mesh, screen, spare part
Se connecter pour consulter vos tarifs contractuels et ceux de votre entreprise/organisme
Le tarif et la disponibilité ne sont pas disponibles actuellement.
A propos de cet article
Code UNSPSC :
41115700
Nomenclature NACRES :
SB.52
Passer à
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
Conditionnement
1 ea of
Catégories apparentées
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 4
Cet article
GE18-1479-01GE28-9356-04GE17-1166-01
packaging
1 ea of
packaging
1 ea of
packaging
pkg of 1 ea
packaging
pkg of 1 ea
Liying Wang et al.
Chemosphere, 245, 125603-125603 (2019-12-20)
Elevated arsenic (As) in groundwater is an urgent environmental problem that has caused serious endemic diseases in Datong basin, China. The fate and toxicity of As are generally regulated by microbial As metabolic processes. However, little is known about the
Gopal Bera et al.
Marine pollution bulletin, 150, 110713-110713 (2019-11-24)
The water-soluble compounds of oil (e.g. low molecular weight PAHs) dissolve as a function of their physicochemical properties and environmental conditions, while the non-soluble compounds exist as dispersed droplets. Both the chemical and physical form of oil will affect the
Young-Wook Chin et al.
Enzyme and microbial technology, 135, 109505-109505 (2020-03-10)
Amylosucrase (ASase) has great industrial potential owing to its multifunctional activities, including transglucosylation, polymerization, and isomerization. In the present study, the properties of Deinococcus geothermalis ASase (DGAS) expressed in Corynebacterium glutamicum (cDGAS) and purified via Ni-NTA affinity chromatography were compared
Vassilia J Sinanoglou et al.
Foods (Basel, Switzerland), 8(12) (2019-12-08)
The implementation of Infrared (IR) radiation in heated greenhouses possesses the advantage of high directional control and focused compensation of energy losses, appropriate for creating local microclimate conditions in highly energy-consuming systems, such as greenhouses. Moreover, it can efficiently maintain
Séverine Allegra et al.
Water research, 172, 115496-115496 (2020-01-24)
The mechanism underlying Legionella aerosolization and entry into the respiratory tract remains poorly documented. In previous studies, we characterized the aerodynamic behaviour of Legionella aerosols and assessed their regional deposition within the respiratory tract using a human-like anatomical model. The