Key Documents
EP5820742003
Buckets for Eppendorf® S-4-104 Rotor
round, capacity 750 mL, pack of 2
About This Item
Code UNSPSC :
41103910
Nomenclature NACRES :
NB.82
Matériaux
round
Conditionnement
pack of 2
Fabricant/nom de marque
Eppendorf® 5820742003 (Global)
Technique(s)
centrifugation: suitable
Capacité
750 mL
Informations légales
Eppendorf is a registered trademark of Eppendorf AG