Key Documents
CLS354014
Corning® Falcon® Sample Container
capacity 110 mL, lid: no, sterile
Synonyme(s) :
Corning container
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.13
Matériaux
polypropylene
Stérilité
sterile
Caractéristiques
lid: no
Fabricant/nom de marque
Corning 354014
Capacité
110 mL
Application
Liquid Handling
Informations légales
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.