Principaux documents
AXYP384120SQC
Corning® Axygen® Deep Well Microplate
size 384 wells, working volume 120 μL, Not Treated, pkg of 10x5plates/cs
Synonyme(s) :
Corning cell culture plate
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux:
clear plate
square bottom clear polypropylene wells
Volume maximum:
130 μL
Taille:
384 wells
Volume de travail:
120 μL
Matériaux
clear plate
square bottom clear polypropylene wells
Stérilité
non-sterile
Caractéristiques
lid: no
skirt
Not Treated
Fabricant/nom de marque
Axygen Inc P-384-120SQ-C
Corning
Conditionnement
pkg of 10x5plates/cs
Volume maximum
130 μL
Taille
384 wells
Volume de travail
120 μL
Type de liaison
non-treated surface
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.