Principaux documents
Z413526
Sigma-Aldrich
Aldrich® three-way adapter
joint: ST/NS 19/22
Synonyme(s) :
3 way adapter, glass adapter, glass adapters
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Joint
joint: ST/NS19/22
Catégories apparentées
Description générale
Side-arm is at 75° angle from bottom joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC