Principaux documents
Z654795
IKA® VORTEX 3 attachments
VG 3.3 universal attachment, 150 mm, with rubber insert (continuous / touch operation), 1/cs
About This Item
Code UNSPSC :
41103814
Nomenclature NACRES :
NB.73
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
IKA 3342400
Diamètre
150 mm
Description générale
IKA VG 3.3 Universal attachment, round, 150mm with rubber insert forcontinuous / touch operation. For use with IKA Vortex 2 and 3 shakers.
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG