Principaux documents
Z304328
Quickfit® dropping funnel
capacity 25 mL, joint: ST/NS 14/23
Synonyme(s) :
D1/01
About This Item
Code UNSPSC :
41121809
Joint
joint: ST/NS14/23
Fabricant/nom de marque
Quickfit D1/01
Disponibilité
not available in USA and Canada
Capacité
25 mL
Description générale
Cylindrical, with ground glass joints and glass stopcock.
Informations légales
Quickfit is a registered trademark of SciLabware Limited