Z261386
Stainless steel 316 syringe needle, pipetting blunt 90° tip
gauge 16, L 6 in.
About This Item
Code UNSPSC :
41122002
eCl@ss :
32070500
Nomenclature NACRES :
NB.03
Produits recommandés
Fabricant/nom de marque
Popper 9873
Longueur
6 in.
Calibre
16
Description générale
For those applications requiring the use of plastic syringes with pipetting needles, these needles are specifically designed for secure, leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. Female Luer hubs and needles are made of 316 SS. Noncontaminating, easy to clean and resistant to corrosion.