Principaux documents
Z256307
Sigma-Aldrich
Wrap-it tie installing tool
Heavy-duty version
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.35
Disponibilité
not available in EU
Description générale
Automatically adjusts wrap-it tie tension to desired position. Tension activated cutting blade trims off excess tie.