Key Documents
Z220094
Sigma-Aldrich
Aldrich® short-path distillation head
Joints: ST/NS 24/40 (2), thermometer joint: ST/NS 10/30
About This Item
Code UNSPSC :
41104812
Nomenclature NACRES :
NB.43
Joint
thermometer joint: ST/NS10/30
Joints: ST/NS24/40 (2)
Profondeur d'immersion
75 mm
Description générale
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC