Principaux documents
Z173428
Sigma-Aldrich
Aldrich® Schlenk-type female joint cap
female joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.43
Étiquette/Marqueur
J
K
Joint
female joint: ST/NS14/20
Description générale
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Liaison
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Certificats d'analyse (COA)
Lot/Batch Number
Désolés, nous n'avons pas de COA pour ce produit disponible en ligne pour le moment.
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.