Documents
Z169064
Coupe-verre diamant
nickel plated, pkg of 1 ea
Synonyme(s) :
Diamond cutter, Glass cutting tool, Precision glass cutter
About This Item
Code UNSPSC :
41122406
Nomenclature NACRES :
NB.35
Matériaux
nickel plated
Conditionnement
pkg of 1 ea
Description générale
Scores/breaks single or double-strength glass. Nickel-plated brass cutter with wooden handle.