Key Documents
Z167576
Sigma-Aldrich
Aldrich® Schlenk-type female joint cap
male joint: ST/NS 14/20
Synonyme(s) :
Aldrich® Schlenk-type hex head stopper
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.43
Étiquette/Marqueur
M
Joint
male joint: ST/NS14/20
Description générale
Thermoplastic screw caps with Viton O-rings usable to 140°C. This stopper has a hexagonal top.
Liaison
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.