Z107476
Aldrich® fritted adapter
female joint: ST/NS 24/40, male joint: ST/NS 24/40
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Joint
female joint: ST/NS24/40
male joint: ST/NS24/40
Description générale
The extra-coarse fritted disc (diameter: 20 mm, porosity A, 145-174 micron) prevents solids from being pulled into the condenser when removing solvents from solid-solvent mixtures. Overall adapter length: 150 mm.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC