Principaux documents
Z107468
Wheaton connecting adapter
To hose connector.
Synonyme(s) :
vacuum adapter, vacuum adapters, wheaton adapters
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Fabricant/nom de marque
WHEATON®
Description générale
13-425 screw-threaded connector.
All wetted parts are PTFE.
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.