Principaux documents
WHA10416230
Whatman® Nytran SuPerCharge (SPC) nylon blotting membranes
W × L 11 cm × 14 cm, pore size 0.45 μm, pkg of 10 ea
Synonyme(s) :
Z698067, blotting membrane, nylone membranes
About This Item
Code UNSPSC :
41105339
Nomenclature NACRES :
NB.22
Matériaux
Nylon membrane (Nytran SuPerCharge (SPC))
Conditionnement
pkg of 10 ea
Fabricant/nom de marque
Whatman 10416230
Whatman Article No. 28415367 (US reference)
Larg. × L
11 cm × 14 cm
Dimension de pores
0.45 μm
0.45 μm
Adéquation
suitable for (Southern and Northern blotting; colony and plaque lifts and Dot-/Slot-Blots)
Description générale
Nylon membrane providing consistently high signal with extremely low background. Nytran SuPerCharge (SPC) Blotting Membranes provide greater reproducibility of results across the membrane and from blot to blot.
Nytran SPC membrane is cast uniformly on both sides of a support matrix, demonstrating excellent symmetry. This gives the membrane the ability to lie flat without curling. With typical manufacturing techniques, increasing positive charge tends to increase background. Nytran SPC membranes are manufactured using a process that allows the combination of high positive charge with low background. Whether using radioactive or nonradioactive detection techniques, Nytran SPC consistently gives high signal with extremely low background. Whatman Nytran nylon membranes are available in two formats: Nytran N is moderately charged and Nytran SuPerCharge (SPC) has a very high positive charge.
Caractéristiques et avantages
  • Very high positive charge.
  • Improvements in the manufacturing process result in a membrane with a higher density of nylon per unit area.
  • The increased charge and greater nylon density provide increased binding sites for your samples.
  • Very uniform pore size and pore distribution compared to typical nylon membranes.
  • Free of surface microvoids, which are common in other membranes.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Nytran is a trademark of Cytiva
Whatman is a registered trademark of Cytiva