TEM-LFH2100AR1.3
QUANTIFOIL on London Finder Grids (H2)
Gold Grids, size 200 mesh, pack of 100
Synonyme(s) :
Electron Microscopy Grids from QUANTIFOIL
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Produits recommandés
Conditionnement
pack of 100
Taille
200 mesh
Description générale
QUANTIFOIL Gold Grids, 200 Mesh, 1.2 micron hole, 2.5 micron period. London Finder Grids (H2), Pitch 127 micron, 200 mesh, 100/pk