Key Documents
TEM-FCF200NI
Sigma-Aldrich
Formvar/Carbon Supported Nickel Grids
size 200 mesh, box of 25 ×
Synonyme(s) :
Formvar/Carbon 200 Mesh, Nickel, 25pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Conditionnement
box of 25 ×
Taille
200 mesh
Comparer avec des articles similaires
Voir la comparaison complète
Montrer les différences
1 of 4
Cet article
ABF32MABF276MABF854
clone
9D5.1, monoclonal
clone
polyclonal
clone
17, monoclonal
clone
6B4, monoclonal
antibody form
purified immunoglobulin
antibody form
affinity isolated antibody
antibody form
purified immunoglobulin
antibody form
purified immunoglobulin
biological source
mouse
biological source
rabbit
biological source
mouse
biological source
mouse
species reactivity
human, mouse
species reactivity
mouse, rat, human, horse
species reactivity
human
species reactivity
human
isotype
IgG1κ
isotype
-
isotype
IgG1κ
isotype
IgG1κ
Description générale
A formvar coated TEM grid, stabilized with evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections.
Thickness: 10nm Formvar and 1nm Carbon
  • Faites votre choix parmi les versions les plus récentes :