Principaux documents
SYNV141005
Synthware conical bottom reaction vial
5 mL, joint: ST/NS 14/20
Synonyme(s) :
Synthware V141005, reaction tube
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware V141005
Capacité
5 mL
Description générale
Heavy wall vials have an internal conical bottom and flat bottom.
Informations légales
Synthware is a trademark of Kemtech America Inc.