Principaux documents
SYNM164380
Synthware solvent transfer manifold
joint: ST/NS 24/40
Synonyme(s) :
Synthware M164380,
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Fabricant/nom de marque
M164380
Catégories apparentées
Description générale
Synthware solvent transfer manifold is used for purifying and transferring solvents under inert conditions. It is supplied with a high vacuum valve featuring a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
Informations légales
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company