Principaux documents
SYNM111005
Synthware Vacuum/Inert Gas Manifold with double bank and high vacuum valves
number of ports, 5, Hose Connections: Front-left-right, Rear-left-right
Synonyme(s) :
Synthware M111005, vacuum manifold, vacuum unit
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
Caractéristiques
number of ports 5
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware M111005
Catégories apparentées
Description générale
General purpose double bank manifold for use with vacuum and inert gas. Supplied with 0-4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and have all banks open. Ports are spaced 100 mm center to center and have an O.D. of 10 mm.
Informations légales
Synthware is a trademark of Kemtech America Inc.