Principaux documents
SYNF603030F
Synthware Buchner funnel
30 mL, porosity: fine
Synonyme(s) :
Synthware F603030F, filtering funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F603030F
Capacité
30 mL
30 mL
Catégories apparentées
Description générale
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Informations légales
Synthware is a trademark of Kemtech America Inc.