Principaux documents
SYNF474250
Synthware Squibb-style separatory funnel with PTFE stopcock and PTFE stopper
250 mL, top joint: ST/NS 24/40
Synonyme(s) :
Synthware F474250, glass separatory funnel
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F474250
Capacité
250 mL
250 mL
Description générale
Supplied with a Glass or PTFE stopper
Informations légales
Synthware is a trademark of Kemtech America Inc.