Principaux documents
SYNF272025C
Synthware fritted gas dispersion tube with 15 degree angle
disc diam. 20 mm, porosity: coarse, stem O.D. × L 8 mm × 150 mm
Synonyme(s) :
Synthware F272025C, glass gas dispersion tube
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Matériaux
colorless tube
glass tube
Stérilité
non-sterile
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F272025C
Diam. du disque
20 mm
Tige, D.E. × L
8 mm × 150 mm
Description générale
The 15° angle permits tube to be inserted into vessels where a large opening is not available. Stem is 8 mm O.D. × 150 mm length, XC stands for extra coarse porosity.
Informations légales
Synthware is a trademark of Kemtech America Inc.