Documents
SYNF234020C
Synthware chromatography column with vacuum connection and fritted disk
joint: ST/NS 24/40, O.D. × L 40 mm × 200 mm, Disc porosity: coarse
Synonyme(s) :
Synthware F234020C, glass chromatography column
About This Item
Code UNSPSC :
23151816
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Qualité
for chromatography
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F234020C
D.E. × L
40 mm × 200 mm
Description générale
Short column with lower 24/40 joint and hose connection for vacuum. All Synthware columns are heavy wall designed to increase safety.
Informations légales
Synthware is a trademark of Kemtech America Inc.