Key Documents
SYNF224250
Synthware four neck round bottom flask with angled side necks
borosilicate glass flask, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonyme(s) :
Synthware F224250, 4-neck reaction flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.51
Matériaux
borosilicate glass flask
four-neck
round bottom
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware F224250
Description générale
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Informations légales
Synthware is a trademark of Kemtech America Inc.