Key Documents
SYNC264175
Synthware coil-style reflux condenser
joint: ST/NS 24/40, hose size 10 mm
Synonyme(s) :
Synthware C264175, coil-type condenser
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware C264175
Taille du tuyau
10 mm
Description générale
Coil style reflux condenser with tightly wrapped coil providing maximum cooling. Excellent for use with volatile solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint. Hose connections have an O.D. of 8mm or 10mm at the middle serration.
Informations légales
Synthware is a trademark of Kemtech America Inc.