SYNC221420
Synthware™ air reflux condenser
joint: ST/NS 14/20, L 96 mm
Synonyme(s) :
Synthware C221420, Glass air condenser
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Produits recommandés
Joint
joint: ST/NS14/20
Conditionnement
pack of 1 ea
Fabricant/nom de marque
C221420
Longueur
96 mm
Description générale
Synthware™ air reflux is a condenser enhancing separation through a reflux mechanism. With upper standard taper outer joint (14/20) to fit various laboratory glassware and a lower drip tip joint. Overall length is 143 mm and length between the joints is 96 mm.
Informations légales
Synthware is a trademark of Kemtech America Inc.