Principaux documents
SYNA282414
Synthware anti-splash adapter
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 14/20
Synonyme(s) :
Synthware A282414, bump trap, rotary evaporator adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS14/20
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A282414
Capacité
100 mL
Catégories apparentées
Description générale
This design prevents possible contamination from rotary evaporator.
Informations légales
Synthware is a trademark of Kemtech America Inc.