Key Documents
DWK217215408
DURAN® narrow mouth reaction flask with beaded rim
glass flask, flask capacity (10,000 mL), single neck, round bottom
Synonyme(s) :
217215408
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.17
Matériaux
glass flask
round bottom
Conditionnement
case of 10 ea
Fabricant/nom de marque
DURAN® 217215408
Contenance du flacon/de la fiole
(10,000 mL)
Description générale
The geometry permits very uniform heating
Caractéristiques et avantages
• Complies with DIN ISO 1773• Manufactured from borosilicate glass 3.3• Fired-on, highly durable white ceramic
Informations légales
DURAN is a registered trademark of DWK Life Sciences