CLS9999182
Corning® phenolic cap with white rubber liner
18-415 thread
Synonyme(s) :
phenolic caps, screw caps
Fabricant/nom de marque
Corning 9999-182
Raccords
18-415 thread
Catégories apparentées
Description générale
These reusable Corning caps are fabricated from specially formulated phenolic resin to resist the effects of temperature and steam in autoclaving. Caps are provided with a glued-in white rubber liner.
Informations légales
Corning is a registered trademark of Corning, Inc.