Principaux documents
BR92763
BRAND® Erlenmeyer flask with beaded rim and graduation, narrow mouth
volume 2000 mL
Synonyme(s) :
Erlenmeyer flasks, Flasks
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
borosilicate glass
flat bottom
Joint
joint neck50 mm
Stérilité
non-sterile
Caractéristiques
without cap
Conditionnement
pack of 10 ea
Fabricant/nom de marque
BRAND 92763
Capacité
2000 mL
Volume
2000 mL
Description générale
The BRAND® Erlenmeyer Flask with beaded rim and graduations is a high-quality laboratory vessel designed for versatile use in chemical and biological applications. Made from Boro 3.3 glass, this flask meets DIN ISO 1773 standards, ensuring durability and reliability in various laboratory environments.
Autres remarques
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG