Principaux documents
BR701934
BRAND® replacement piston rod for TRANSFERPETTOR
for fixed and digital type 10-50 and 50 μL
Synonyme(s) :
Accessories, Pipettes, Piston rod, Piston-operated pipettes, Positive displacement principle, Transferpettor
About This Item
Code UNSPSC :
41121501
Nomenclature NACRES :
NB.01
Matériaux
stainless steel
Conditionnement
pack of 3 ea
Fabricant/nom de marque
BRAND 701934
Description générale
The BRAND® Replacement Piston Rod for the TRANSFERPETTOR is designed to ensure optimal performance and precision in liquid handling. Starting from 20 µl, product comes equipped with a seal. The replacement rod is easy to install and maintain, providing a reliable solution for users seeking to extend the life of their TRANSFERPETTOR while maintaining accuracy and efficiency in their pipetting tasks.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG