BAH420530000
SP Bel-Art® Dry-Keeper Small, Stacking Desiccator Cabinet
polystyrene, volume 0.14 ft3
About This Item
Code UNSPSC :
41123002
Nomenclature NACRES :
NB.43
Produits recommandés
Matériaux
polystyrene
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Bel-Art H42053-0000
Volume
0.14 ft3
Description générale
Small, clear polystyrene desiccator cabinet safely stacks up to 5 units high for maximum use of dry storage space.
- Pull down door slides out of the way into a compartment located underneath the cabinet
- Ready to use jar of silica gel is included
Informations légales
Bel-Art is a registered trademark of SP Scienceware, a division of SP