Principaux documents
BAH131852430
Clavies® autoclave bags
width × H × thickness 24 in. (610 mm) × 30 in. (760 mm) × 2 mil (0.05 mm), clear, pack of 100 ea
Synonyme(s) :
sterilzation bag, transparent autoclave bags
About This Item
Code UNSPSC :
41122800
Nomenclature NACRES :
NB.35
Matériaux
clear
Conditionnement
pack of 100 ea
Fabricant/nom de marque
Bel-Art H13185-2430
width × H × thickness
24 in. (610 mm) × 30 in. (760 mm) × 2 mil (0.05 mm)
Catégories apparentées
Description générale
These transparent autoclave bags are made of 0.05mm (2 mil) thick polypropylene for discarding used Petri dishes, membrane filters, multi-well cell culture plates, cell culture flasks, culture plates, pipettes, and more.
Caractéristiques et avantages
  • Three largest sizes hold 65, 200 and 240 standard Petri dishes respectively
  • Steam autoclavable at 135°C (275°F)
Informations légales
Clavies is a registered trademark of SP Scienceware, a division of SP