Merck
  • Mechanistic insights into the reactions of hydride transfer versus hydrogen atom transfer by a trans-dioxoruthenium(VI) complex.
Mechanistic insights into the reactions of hydride transfer versus hydrogen atom transfer by a trans-dioxoruthenium(VI) complex.
Dalton transactions (Cambridge, England : 2003) (2015-03-27)
Sunder N Dhuri, Yong-Min Lee, Mi Sook Seo, Jaeheung Cho, Dattaprasad D Narulkar, Shunichi Fukuzumi, Wonwoo Nam
ABSTRACT
A mononuclear high-valent trans-dioxoruthenium(VI) complex, trans-[Ru(VI)(TMC)(O)2](2+) (TMC = 1,4,8,11-tetramethyl-1,4,8,11-tetraazacyclotetradecane), was synthesized and characterized by various spectroscopic techniques and X-ray crystallography. The reactivity of the trans-[Ru(VI)(TMC)(O)2](2+) complex was investigated in hydride transfer and hydrogen atom transfer reactions. The mechanism of hydride transfer from dihydronicotinamide adenine dinucleotide (NADH) analogues to trans-[Ru(VI)(TMC)(O)2](2+), which proceeds via a proton-coupled electron transfer (PCET), followed by a rapid electron transfer (ET), has been proposed by the observation of a good linear correlation between the log rate constants of trans-[Ru(VI)(TMC)(O)2](2+) and p-chloranil (Cl4Q) and a large kinetic isotope effect (KIE) value of 13(1). In the case of the oxidation of alkyl hydrocarbons by the trans-[Ru(VI)(TMC)(O)2](2+) complex, the second-order rate constants were dependent on the C-H bond dissociation energy (BDE) of the substrates, and a large KIE value of 26(2) was obtained in the oxidation of xanthene and deuterated xanthene-d2 by the trans-[Ru(VI)(TMC)(O)2](2+) complex, indicating that the C-H bond activation of alkyl hydrocarbons proceeds via an H-atom abstraction in the rate-determining step.
Product Description
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
1,4-Cyclohexadiene, 97%
Sigma-Aldrich
9,10-Dihydroanthracene, 97%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen Peroxide Solution, 30% (w/w), puriss. p.a., reag. ISO, reag. Ph. Eur.
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, purum p.a., ≥35% (RT)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Potassium aquapentachlororuthenate(III)