Merck
  • A diet enriched with the omega-3 fatty acid docosahexaenoic acid reduces amyloid burden in an aged Alzheimer mouse model.
A diet enriched with the omega-3 fatty acid docosahexaenoic acid reduces amyloid burden in an aged Alzheimer mouse model.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2005-03-25)
Giselle P Lim, Frédéric Calon, Takashi Morihara, Fusheng Yang, Bruce Teter, Oliver Ubeda, Norman Salem, Sally A Frautschy, Greg M Cole
ABSTRACT
Epidemiological studies suggest that increased intake of the omega-3 (n-3) polyunsaturated fatty acid (PUFA) docosahexaenoic acid (DHA) is associated with reduced risk of Alzheimer's disease (AD). DHA levels are lower in serum and brains of AD patients, which could result from low dietary intake and/or PUFA oxidation. Because effects of DHA on Alzheimer pathogenesis, particularly on amyloidosis, are unknown, we used the APPsw (Tg2576) transgenic mouse model to evaluate the impact of dietary DHA on amyloid precursor protein (APP) processing and amyloid burden. Aged animals (17-19 months old) were placed in one of three groups until 22.5 months of age: control (0.09% DHA), low-DHA (0%), or high-DHA (0.6%) chow. beta-Amyloid (Abeta) ELISA of the detergent-insoluble extract of cortical homogenates showed that DHA-enriched diets significantly reduced total Abeta by >70% when compared with low-DHA or control chow diets. Dietary DHA also decreased Abeta42 levels below those seen with control chow. Image analysis of brain sections with an antibody against Abeta (amino acids 1-13) revealed that overall plaque burden was significantly reduced by 40.3%, with the largest reductions (40-50%) in the hippocampus and parietal cortex. DHA modulated APP processing by decreasing both alpha- and beta-APP C-terminal fragment products and full-length APP. BACE1 (beta-secretase activity of the beta-site APP-cleaving enzyme), ApoE (apolipoprotein E), and transthyretin gene expression were unchanged with the high-DHA diet. Together, these results suggest that dietary DHA could be protective against beta-amyloid production, accumulation, and potential downstream toxicity.
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal (751-770) Rabbit pAb, liquid, Calbiochem®