Key Documents
Z740503
smartSlicer Gel Cutting Tool & RaZor
smartSlicer™ Gel Cutting Tool & RaZor
standard
Synonym(s):
electrophoresis gel cutter, gel knife, gel razor, gel razor replacement, safety cutter, safety razor, western gel cutting
About This Item
UNSPSC Code:
41105300
NACRES:
NB.35
Pricing and availability is not currently available.
packaging
pack of 100 ea
manufacturer/tradename
LevGo Inc 30202
L × H × thickness
25 mm × 19 mm × 0.27 mm
General description
At just 0.27 mm (0.011 in.) the US Patent Pending smartSlicer is as thin as a single edge industrial razor blade but much safer - the smartSlicer is unlikely to slice you!
  • The smartSlicer is able to cleanly slice gels* right on the light Box without scratching the surface of the box
  • The smartSlicer can also be used to chop, mince or slice some soft tissue samples
  • The smartSlicer is also adjustable - simply cut it with standard scissors to the width you require
Available in two sizes: our Standard size is the same size as an industrial single edge razor. Our Large smartSlicer is 30% larger for bigger jobs.
smartSlicers are made in the USA of food grade PET and packed 100 per pack.
Lab Safety Managers love the smartSlicer!
*Best for 1% to 2% gels.
Legal Information
smartSlicer is a trademark of LevGo Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Active Filters
  1. 1 Ratings-Only Review