Key Documents
Z332798
BRAND® adapter for 25 & 50 mL PD-Tips
non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
50 mL
sterility:
non-sterile
packaging:
pack of 10 ea
manufacturer/tradename:
BRAND 702398
material
polypropylene
sterility
non-sterile
packaging
pack of 10 ea
manufacturer/tradename
BRAND 702398
maximum volume
50 mL
General description
The PD-Tips II dispenser tips for efficient, step-by-step dispensing enhance the proven features of the classic BRAND PD-Tips.Thanks to patented size coding, HandyStep® touch and HandyStep® electronic from BRAND automatically detect the tip volume. The tips are also compatible with all other instruments of the HandyStep® family and with repetitive pipettes from other manufacturers.The positive displacement principle enables precision work, even with viscous media or media with high vapor pressure. With tightly sealed pistons, the PD-Tips II effectively protect the stepper from aerosols. This prevents contamination and ensures long service life of the instrument.At a glance- Volumes from 0.1 ml to 50 ml- Compatible with different repetitive pipets- Compatible with automatic size detection: HandyStep® touch, HandyStep® electronic, Gilson® Repetman, Rainin® AutoRep E- Compatible without automatic size detection: HandyStep® S, HandyStep®, Rainin® AutoRep S, Rainin® AutoRep M, Eppendorf® Multipette® 4780 and EDOS® 5221 (the above-mentioned brands are property of their respective owners)- Cleanroom quality- Comply with the requirements of ISO 8655- Delivered with batch certificate- Manufactured from high-quality raw materials (cylinder: PP / Piston: PE-HD, 0.1 ml: LCP)- Adapter for sizes 25 ml and 50 ml included- PD-Tips II cannot be autoclaved; the adapters of the 25ml and 50ml sizes can be autoclaved at 121° C.Adapters for size 25 and 50 ml PD-Tips, PP, autoclavable. Non-sterile or sterile in BIO-CERT® quality.CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
Other Notes
BR702398 (India)
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
EDOS is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Gilson is a registered trademark of Gilson, Inc.
HandyStep is a registered trademark of BRAND GMBH + CO KG
Multipette is a registered trademark of Eppendorf AG
Rainin is a registered trademark of Rainin Instrument, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)