Skip to Content
Merck
All Photos(1)

Key Documents

SML3722

Sigma-Aldrich

Dinaciclib

≥98% (HPLC)

Synonym(s):

(2S)-1-[3-Ethyl-7-[[(1-oxido-3-pyridinyl)methyl]amino]pyrazolo[1,5-a]pyrimidin-5-yl]-2-piperidineethanol, 2-[(2S)-1-[3-Ethyl-7-[(1-oxidopyridin-1-ium-3-yl)methylamino]pyrazolo[1,5-a]pyrimidin-5-yl]piperidin-2-yl]ethanol, MK 7965, MK-7965, MK7965, SCH 727965, SCH-727965, SCH727965

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28N6O2
CAS Number:
Molecular Weight:
396.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

[n+]1(cc(ccc1)CNC2=CC(=Nc4[n]2ncc4CC)N3[C@@H](CCCC3)CCO)[O-]

InChI key

PIMQWRZWLQKKBJ-SFHVURJKSA-N

Biochem/physiol Actions

Dinaciclib (SCH 727965) is a potent and selective cyclin-dependent kinase (CDK) inhibitor (CDK1/2/5/6/7/9 IC50 = 3/1/1/12/7/4 nM; Little or no inhibition of 41 other kinases at 10 μM) with potent antiproliferation activity in A2780 ovarian cancer cultures (IC50 = 4 nM by thymidine uptake assays). Dinaciclib exhibits potent anti-cancer efficacy in cultures (IC50 = 6-17 nM against 14 cancer cell lines) and in an A2780 tumor xenograft model in mice in vivo (8-48 mg/kg via daily i.p.).
Potent and selective cyclin-dependent kinase (CDK) inhibitor with potent anti-cancer efficacy in cultures and in mice in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service