Skip to Content
Merck
All Photos(1)

Key Documents

SML1360

Sigma-Aldrich

Cariporide

≥98% (HPLC)

Synonym(s):

HOE-642, N-(Aminoiminomethyl)-4-(1-methylethyl)-3-(methylsulfonyl)-benzamide, N-(Diaminomethylene)-4-isopropyl-3-(methylsulfonyl)benzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17N3O3S
CAS Number:
Molecular Weight:
283.35
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(NC(C1=CC(S(C)(=O)=O)=C(C(C)C)C=C1)=O)=N

InChI

1S/C12H17N3O3S/c1-7(2)9-5-4-8(11(16)15-12(13)14)6-10(9)19(3,17)18/h4-7H,1-3H3,(H4,13,14,15,16)

InChI key

IWXNYAIICFKCTM-UHFFFAOYSA-N

Biochem/physiol Actions

Cariporide is a selective inhibitor of the Na+/H+ exchanger subtype 1 (NHE-1), also known as the Na+/H+ antiporter. Cariporide has shown cardioprotective and antiarrhythmic effects, and has recently been investigated for anticancer activity.
Cariporide is a selective inhibitor of the Na+/H+ exchanger subtype 1 (NHE-1).
Cariporide, a sodium-hydrogen exchange inhibitor[1] can decrease the liberation of myoglobin at the time of ischemia/reperfusion and reduces the liberation of NE (norepinephrine) during ischemia.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuoqun Fang et al.
Shock (Augusta, Ga.), 54(6), 819-827 (2020-06-05)
Na/H exchanger 1 (NHE1) is a ubiquitously expressed protein on mammalian plasma membranes and involved in cell apoptosis and tissue injury. Our previous study found that NHE1 inhibition prevents burn-induced acute lung injury (ALI). However, the potential mechanism of NHE1
Jair C Kwan et al.
Heart, lung & circulation, 24(3), 306-318 (2014-12-03)
Storage of donor hearts in cardioplegic solutions supplemented with agents that mimic the ischaemic preconditioning response enhanced their post-reperfusion function. The present study examines the minimisation of cell death and activation of pro-survival signalling directed towards maintenance of mitochondrial homeostasis
Shigeru Sakurai et al.
Life sciences, 114(2), 102-106 (2014-08-21)
To examine the effects of cariporide, a Na(+)/H(+) exchanger-1 inhibitor, on cardiac norepinephrine (NE) and myoglobin release during myocardial ischemia/reperfusion by applying a microdialysis technique to the rabbit heart. In anesthetized rabbits, two dialysis probes were implanted into the left

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service