Skip to Content
Merck
All Photos(1)

Key Documents

O3389

Sigma-Aldrich

Anti-Osteopontin antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-OPN

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human

technique(s)

immunohistochemistry: 5-10 μg/mL
neutralization: suitable
western blot: 0.1-0.2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPP1(6696)

Immunogen

purified recombinant human osteopontin.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luisa Piva et al.
The Journal of pathology, 228(2), 251-259 (2012-03-21)
A polymorphism (rs28357094) in the promoter region of the SPP1 gene coding for osteopontin (OPN) is a strong determinant of disease severity in Duchenne muscular dystrophy (DMD). The rare G allele of rs28357094 alters gene promoter function and reduces mRNA
Sara Vianello et al.
Human molecular genetics, 26(17), 3342-3351 (2017-06-09)
Glucocorticoids are beneficial in Duchenne muscular dystrophy (DMD). Osteopontin (OPN), the protein product of SPP1, plays a role in DMD pathology modulating muscle inflammation and regeneration. A polymorphism in the SPP1 promoter (rs28357094) has been recognized as a genetic modifier

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service