Skip to Content
Merck
All Photos(1)

Key Documents

N1000

General Purpose Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified

viscosity 2610 mPa.s (25 °C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

viscosity

2610 mPa.s(25 °C)

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

Please note that the value stated on this page is nominal. All certified values for the current lots of all Paragon General Viscosity Standards can be found on the Paragon Scientific webpage
This product is a certified reference material (CRM) certified to International Standards BS EN ISO / IEC 17025 and ISO 17034 under UKAS accreditation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erica Rinehart et al.
Pathogens (Basel, Switzerland), 7(3) (2018-07-04)
Propionate is a common food preservative and one of the major fermentation acids in the intestines. Therefore, exposure to propionate is frequent for foodborne pathogens and likely takes place under suboxic conditions. However, it is not clear whether the absence
Christian O Dimkpa et al.
The Science of the total environment, 665, 606-616 (2019-02-19)
Plant response to microelements exposure can be modulated based on particle size. However, studies are lacking on the roles of particle size and specific microelements in mixed exposure systems designed for plant nutrition, rather than toxicology. Here, an addition-omission strategy
Hala Elnakat Thomas et al.
Cell reports, 24(9), 2404-2417 (2018-08-30)
Cells adapt to nutrient and energy deprivation by inducing autophagy, which is regulated by the mammalian target of rapamycin (mTOR) and AMP-activated protein kinases (AMPKs). We found that cell metabolism significantly influences the ability to induce autophagy, with mitochondrial complex

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service