GBL845237
Grace Bio-Labs ProCrystal™ protein crystallization cover
cut per individual well, plastic liner, wells, 96
Synonym(s):
adhesive crystallization cover, precut paper frame crystallization cover
All Photos(2)
About This Item
UNSPSC Code:
41122410
NACRES:
NB.15
Recommended Products
material
plastic film
sterility
non-sterile
feature
wells 96
packaging
pack of 5 ea
manufacturer/tradename
Grace Bio-Labs 845237
W × L
78 mm × 120 mm
Related Categories
General description
ProCrystal protein crystallization covers are designed for high throughput protein x-ray crystallography in 96-well plates. The ProCrystal material is extremely hydrophobic to maintain drop footprint and segregation, even with protocols using MPD, glycerol or detergents. Grace Bio-Labs has collaborated with leading crystallographers to develop our ProCrystal covers using materials with the highest optical quality and stringent manufacturing processes to ensure clean, particle-free covers.
Features Include:
ProCrystal protein crystallization covers are extremely hydrophobic and UV compatible. Covers accommodate up to 3 protein drops per well, and are available as frame or frameless, with either paper or plastic liners, and in two cut formats: pre-cut or uncut. Our protein crystallization covers are compatible with SBS-compliant microtiter plates and TTP Mosquito Instruments. The ProCrystal cover is also UV compatible with the highest optical quality available. The stringent manufacturing process and quality control ensure clean, particle-free material shipped ready to use. ProCrystal protein crystallization covers are available in an uncut, or pre-cut format that allows removal of covers from individual wells at different times.
Features Include:
- TTP Labtech Mosquito compatible
- Extremely hydrophobic
- Ready-to-use, dust-free
- UV compatible
- No noticeable x-ray diffraction
- Accommodates up to 3 protein drops per well
- Two formats: pre-cut or uncut
ProCrystal protein crystallization covers are extremely hydrophobic and UV compatible. Covers accommodate up to 3 protein drops per well, and are available as frame or frameless, with either paper or plastic liners, and in two cut formats: pre-cut or uncut. Our protein crystallization covers are compatible with SBS-compliant microtiter plates and TTP Mosquito Instruments. The ProCrystal cover is also UV compatible with the highest optical quality available. The stringent manufacturing process and quality control ensure clean, particle-free material shipped ready to use. ProCrystal protein crystallization covers are available in an uncut, or pre-cut format that allows removal of covers from individual wells at different times.
Legal Information
ProCrystal is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)