Skip to Content
Merck
All Photos(1)

Key Documents

G9887

Sigma-Aldrich

γ-Globulins from sheep

≥99% (agarose gel electrophoresis)

Synonym(s):

Gamma globulin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

sheep

Quality Level

Assay

≥99% (agarose gel electrophoresis)

form

powder

technique(s)

cell culture | mammalian: suitable

impurities

≤4% NaCl

storage temp.

−20°C

General description

γ-Globulin molecules is made up of four polypeptide chains, of which two have molecular weights of about 50,000 and two of about 20,000. These chains are cross-linked by disulfide bonds.

Application

γ-Globulins from sheep were used to study the influence of HDL-cholesterol-elevating drugs on the in vitro activity of the HDL receptor SR-BI. It was used in a study to test the interstitial transport of rabbit and sheep antibodies in normal and neoplastic tissues. It was also used in a study to test the enhanced progesterone and testosterone secretion, and depressed estradiol secretion in vitro from small white follicle cells of incubating turkey hens.

Preparation Note

Prepared from Cohn Fraction II, III

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T E Porter et al.
General and comparative endocrinology, 74(3), 400-405 (1989-06-01)
This study was conducted to determine if lower steroid secretion by the small white follicles in incubating turkey hens contributes to lower circulating steroid concentrations during this time. Turkey hens were grouped as either laying or incubating. Serum samples and
M A Clauss et al.
Cancer research, 50(12), 3487-3492 (1990-06-15)
Interstitial transport of fluorescein isothiocyanate-conjugated nonspecific polyclonal rabbit and sheep IgG was studied in normal (mature granulation) and neoplastic (VX2 carcinoma) tissues grown in a rabbit ear chamber. The interstitial concentration gradients after i.v. injection were analyzed to yield effective
[Postoperative thrombosis prevention using low-dose heparin].
A E Schaer et al.
Archiv fur Gynakologie, 219(1-4), 545-545 (1975-11-18)
Thomas J F Nieland et al.
Journal of lipid research, 48(8), 1832-1845 (2007-05-30)
Treatment of atherosclerotic disease often focuses on reducing plasma LDL-cholesterol or increasing plasma HDL-cholesterol. We examined in vitro the effects on HDL receptor [scavenger receptor class B type I (SR-BI)] activity of three classes of clinical and experimental plasma HDL-cholesterol-elevating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service